missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-5-methoxyphenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-4228513
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
89694-46-2
C7H8BClO3
186.40
MFCD06659858
REFXAANPQCJZRY-UHFFFAOYSA-N
2-chloro-5-methoxyphenyl boronic acid, 2-chloro-5-methoxyphenylboronicacid, 3-borono-4-chloroanisole, 2-chloro-5-methoxybenzeneboronic acid, 2-chloro-5-methoxy-phenyl boronic acid, boronic acid, 2-chloro-5-methoxyphenyl, pubchem6932, acmc-209r2h, 2-chloro-5-methoxylphenylboronic acid
17750233
(2-chloro-5-methoxyphenyl)boronic acid
COC1=CC=C(Cl)C(=C1)B(O)O
This item is not returnable. View return policy
89694-46-2
C7H8BClO3
186.40
MFCD06659858
REFXAANPQCJZRY-UHFFFAOYSA-N
2-chloro-5-methoxyphenyl boronic acid, 2-chloro-5-methoxyphenylboronicacid, 3-borono-4-chloroanisole, 2-chloro-5-methoxybenzeneboronic acid, 2-chloro-5-methoxy-phenyl boronic acid, boronic acid, 2-chloro-5-methoxyphenyl, pubchem6932, acmc-209r2h, 2-chloro-5-methoxylphenylboronic acid
17750233
(2-chloro-5-methoxyphenyl)boronic acid
COC1=CC=C(Cl)C(=C1)B(O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 111.0°C to 114.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 186.4
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-chloro-5-methoxyphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-5-methoxyphenylboronic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'