missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-iodopyridine, 98+%, Thermo Scientific Chemicals

Catalog Number 10649194
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
69045-79-0
C5H3ClIN
239.44
QWLGCWXSNYKKDO-UHFFFAOYSA-N
2-chloro-5-indopyridine, 2-chloro-5-iodo-pyridine, pyridine, 2-chloro-5-iodo, 2-chloro-5-iodo pyridine, zlchem 492, pubchem2078, acmc-1bafa, 2-chloro-5 iodopyridine, 2-chloro-5-iodopyridine, 5-iodo-2-chloro-pyridine
4397176
2-chloro-5-iodopyridine
C1=CC(=NC=C1I)Cl
This item is not returnable. View return policy
69045-79-0
C5H3ClIN
239.44
QWLGCWXSNYKKDO-UHFFFAOYSA-N
2-chloro-5-indopyridine, 2-chloro-5-iodo-pyridine, pyridine, 2-chloro-5-iodo, 2-chloro-5-iodo pyridine, zlchem 492, pubchem2078, acmc-1bafa, 2-chloro-5 iodopyridine, 2-chloro-5-iodopyridine, 5-iodo-2-chloro-pyridine
4397176
2-chloro-5-iodopyridine
C1=CC(=NC=C1I)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 96.0°C to 99.0°C
Color Beige-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 239.44
Percent Purity 98+%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 2-Chloro-5-iodopyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-5-iodopyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'