missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-iodo-3-(trifluoromethyl)pyridine, 95%, Thermo Scientific Chemicals

Catalog Number 15458846
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
887707-25-7
C6H2ClF3IN
307.44
MFCD12024302
KUASPTUGCBGXAE-UHFFFAOYSA-N
2-chloro-5-iodo-3-trifluoromethyl pyridine, 2-chloro-3-trifluoromethyl-5-iodo-pyridine, 2-chloro-3-trifluoromethyl-5-iodopyridine, 2-choro-5-iodo-3-trifluoromethylpyridine, 2-chloro-3-trifluoromethyl-5-iodo pyridine
11500513
2-chloro-5-iodo-3-(trifluoromethyl)pyridine
FC(F)(F)C1=CC(I)=CN=C1Cl
This item is not returnable. View return policy
887707-25-7
C6H2ClF3IN
307.44
MFCD12024302
KUASPTUGCBGXAE-UHFFFAOYSA-N
2-chloro-5-iodo-3-trifluoromethyl pyridine, 2-chloro-3-trifluoromethyl-5-iodo-pyridine, 2-chloro-3-trifluoromethyl-5-iodopyridine, 2-choro-5-iodo-3-trifluoromethylpyridine, 2-chloro-3-trifluoromethyl-5-iodo pyridine
11500513
2-chloro-5-iodo-3-(trifluoromethyl)pyridine
FC(F)(F)C1=CC(I)=CN=C1Cl

It is used as pharmaceutical intermediate and as an advanced chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate and as an advanced chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from exposure to light.
TRUSTED_SUSTAINABILITY
Melting Point 43°C
Quantity 250 mg
Beilstein 10249610
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 307.44
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Chloro-5-iodo-3-(trifluoromethyl)pyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'