missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-5-fluoropyrimidine, 98%, Thermo Scientific Chemicals

Catalog Number p-4090008
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
62802-42-0
C4H2ClFN2
132.53
MFCD03788197
AGYUQBNABXVWMS-UHFFFAOYSA-N
2-chloro-5-fluoro-pyrimidine, 5-fluoro-2-chloropyrimidine, pyrimidine, 2-chloro-5-fluoro, 2-chlor-5-fluorpyrimidin, 2-chloro-5-fluoro pyrimidine, pubchem4684, acmc-1b6zl, ksc352s4h, #, 2-chloro-5-fluoro-1,3-diazine
583466
2-chloro-5-fluoropyrimidine
C1=C(C=NC(=N1)Cl)F
This item is not returnable. View return policy
62802-42-0
C4H2ClFN2
132.53
MFCD03788197
AGYUQBNABXVWMS-UHFFFAOYSA-N
2-chloro-5-fluoro-pyrimidine, 5-fluoro-2-chloropyrimidine, pyrimidine, 2-chloro-5-fluoro, 2-chlor-5-fluorpyrimidin, 2-chloro-5-fluoro pyrimidine, pubchem4684, acmc-1b6zl, ksc352s4h, #, 2-chloro-5-fluoro-1,3-diazine
583466
2-chloro-5-fluoropyrimidine
C1=C(C=NC(=N1)Cl)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 149°C to 162°C (1013.0 hPa)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 250 mg
Formula Weight 132.53
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-chloro-5-fluoropyrimidine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chloro-5-fluoropyrimidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'