missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-5-fluorobenzyl bromide, 98%, Thermo Scientific™

Catalog Number p-7053665
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
81778-09-8
C7H5BrClF
223.469
MFCD01631419
AUVLFQDKJFSFIX-UHFFFAOYSA-N
2-chloro-5-fluorobenzyl bromide, 2-bromomethyl-1-chloro-4-fluorobenzene, 2-chloro-5-fluorobenzylbromide, 1-bromomethyl-2-chloro-5-fluorobenzene, pubchem7498, acmc-1blqd, 2-bromomethyl-1-chloro-4-fluoro-benzene, 2-chloro5-fluorobenzyl bromide, 2-chloro-5-fluoro-benzylbromide, 2-chloro-5-fluoro-benzyl bromide
2773623
2-(bromomethyl)-1-chloro-4-fluorobenzene
C1=CC(=C(C=C1F)CBr)Cl
This item is not returnable. View return policy
81778-09-8
C7H5BrClF
223.469
MFCD01631419
AUVLFQDKJFSFIX-UHFFFAOYSA-N
2-chloro-5-fluorobenzyl bromide, 2-bromomethyl-1-chloro-4-fluorobenzene, 2-chloro-5-fluorobenzylbromide, 1-bromomethyl-2-chloro-5-fluorobenzene, pubchem7498, acmc-1blqd, 2-bromomethyl-1-chloro-4-fluoro-benzene, 2-chloro5-fluorobenzyl bromide, 2-chloro-5-fluoro-benzylbromide, 2-chloro-5-fluoro-benzyl bromide
2773623
2-(bromomethyl)-1-chloro-4-fluorobenzene
C1=CC(=C(C=C1F)CBr)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.654 g/mL
Boiling Point 65°C to 66°C
Flash Point 17°C (63°F)
Refractive Index 1.566
Quantity 1 g
UN Number UN2924
Formula Weight 200.01
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Chloro-5-fluorobenzyl bromide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'