missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-5-fluoro-3-methylpyridine, 95%

Brand:  Alfa Aesar™ H61447.03

Code : NEW

Additional Details : CAS Number : 38186-84-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15497988

  • £118.83 / 1g


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-5-fluoro-3-methylpyridine
38186-84-4
145.56
MFCD04972725

Specifications

2-Chloro-5-fluoro-3-methylpyridine
38186-84-4
95%
MFCD04972725
1g
Solid
145.56
Videos
SDS