Learn More
2-chloro-5-(chloromethyl)thiophene, 97%, ACROS Organics™
Brand: Acros Organics 221350010
Code : 29
Additional Details : CAS Number : 23784-96-5 Weight : 0.05100kg Transport : UN number : 3265 Chem class : 1a Pack group : II
Quantity | 1g |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
2-chloro-5-(chloromethyl)thiophene | |
96.0 | |
97% | |
167.06 |
23784-96-5 | |
100.0 | |
C5H4Cl2S | |
MFCD00009764 |
Specifications
2-chloro-5-(chloromethyl)thiophene | |
1g | |
96.0 | |
Glass bottle | |
C5H4Cl2S | |
MFCD00009764 |
1.38g/mL | |
23784-96-5 | |
100.0 | |
97% | |
167.06 | |
17, III, 271 |