missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-5-(chloromethyl)thiophene, 97%

Brand:  Alfa Aesar™ H54361.03

Code : NEW

Additional Details : CAS Number : 23784-96-5 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15450678

  • £30.42 / 1g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-5-(chloromethyl)thiophene
23784-96-5
167.06
MFCD00009764

Specifications

2-Chloro-5-(chloromethyl)thiophene
83° to 85°C
1g
1.575
97%
MFCD00009764
1.385g/mL
Colorless to brown
23784-96-5
Liquid
167.06
Videos
SDS