missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-5-(2,5-dichlorophenylcarbamoyl)benzeneboronic acid, 97%

Brand:  Alfa Aesar™ H54433.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15401186

  • £395.00 / 1g

Estimated Shipment: 23-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1449132-57-3
344.379
LGHQYAUSSYHBTK-UHFFFAOYSA-N
73995808
B(C1=C(C=CC(=C1)C(=O)NC2=C(C=CC(=C2)Cl)Cl)Cl)(O)O
C13H9BCl3NO3
MFCD22125068
2-chloro-5-2,5-dichlorophenylcarbamoyl benzeneboronic acid, 2-chloro-5-2,5-dichlorophenyl carbamoyl phenylboronic acid, 2-chloro-5-2,5-dichlorophenylcarbamoyl phenylboronic acid
[2-chloro-5-[(2,5-dichlorophenyl)carbamoyl]phenyl]boronic acid

Specifications

2-Chloro-5-(2,5-dichlorophenylcarbamoyl)benzeneboronic acid
1449132-57-3
MFCD22125068
LGHQYAUSSYHBTK-UHFFFAOYSA-N
[2-chloro-5-[(2,5-dichlorophenyl)carbamoyl]phenyl]boronic acid
73995808
97%
1g
C13H9BCl3NO3
2-chloro-5-2,5-dichlorophenylcarbamoyl benzeneboronic acid, 2-chloro-5-2,5-dichlorophenyl carbamoyl phenylboronic acid, 2-chloro-5-2,5-dichlorophenylcarbamoyl phenylboronic acid
B(C1=C(C=CC(=C1)C(=O)NC2=C(C=CC(=C2)Cl)Cl)Cl)(O)O
344.379
344.39
Videos
SDS Documents
Certificates