missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-(trifluoromethyl)phenylacetic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
CAS | 601513-26-2 |
Melting Point | 108°C to 111°C |
Color | White |
Molecular Formula | C9H6ClF3O2 |
MDL Number | MFCD13194288 |
Quantity | 250 mg |
Synonym | 2-2-chloro-4-trifluoromethyl phenyl acetic acid, 2-chloro-4-trifluoromethylphenylacetic acid, 2-chloro-4-trifluoromethyl phenylacetic acid, 2-chloro-4-trifluoromethyl phenyl acetic acid, 2-chloro-4-trifluoromethylphenyl-acetic acid, 2-chloro-4-trifluoromethyl-benzeneacetic acid, benzeneacetic acid, 2-chloro-4-trifluoromethyl |
InChI Key | JLULPIXGVZSEES-UHFFFAOYSA-N |
SMILES | OC(=O)CC1=C(Cl)C=C(C=C1)C(F)(F)F |
IUPAC Name | 2-[2-chloro-4-(trifluoromethyl)phenyl]acetic acid |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'