missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-chloro-4-thien-2-ylpyrimidine, Thermo Scientific™, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 2-chloro-4-thiophen-2-yl pyrimidine, 2-chloro-4-thien-2-ylpyrimidine, pyrimidine, 2-chloro-4-2-thienyl, 2-chloro-4-thiophen-2-yl-pyrimidine, 2-chloro-4-2-thienyl pyrimidine, 2-chloro-4-thien-2-yl pyrimidine, 2-2-chloropyrimidin-4-yl thiophene, pyrimidine,2-chloro-4-2-thienyl |
Formula Weight | 196.66 |
Percent Purity | 97% |
Chemical Name or Material | 2-Chloro-4-thien-2-ylpyrimidine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'