missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-picoline, 98%, ACROS Organics™

Brand:  Acros Organics 368841000

Code : 29

Additional Details : CAS Number : 3678-62-4 Weight : 0.16510kg


Quantity 100mL
Packaging Glass bottle

 View more versions of this product

Product Code. 10581154

  • £381.70 / 100mL


EU Stock 6
Add to basket


Specifications

Chemical Identifiers

2-Chloro-4-picoline
97.5
≥97.5%
C6H6ClN
MFCD00023418
3678-62-4
100.0
98%
127.57
2-Chloro-4-methylpyridine

Specifications

2-Chloro-4-picoline
Clear colorless to light yellow
3678-62-4
100.0
Glass bottle
Liquid
98%
127.57
2-Chloro-4-methylpyridine
1.1510g/mL
100mL
97.5
Authentic
1.5280 to 1.5300 (20°C, 589nm)
≥97.5%
C6H6ClN
MFCD00023418
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Chloro-4-picoline

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS