missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitrophenyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog Number p-7045484
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
23165-64-2
C7H3ClN2O2S
214.62
MFCD00041244
JSYNALWWKRNZDX-UHFFFAOYSA-N
2-chloro-4-nitrophenyl isothiocyanate, 2-chloro-4-nitrophenylisothiocyanate, benzene,2-chloro-1-isothiocyanato-4-nitro, 2-chloro-4-nitrobenzenisothiocyanate, acmc-1cg9c, 2-chloranyl-1-isothiocyanato-4-nitro-benzene
2757736
2-chloro-1-isothiocyanato-4-nitrobenzene
[O-][N+](=O)C1=CC(Cl)=C(C=C1)N=C=S
This item is not returnable. View return policy
23165-64-2
C7H3ClN2O2S
214.62
MFCD00041244
JSYNALWWKRNZDX-UHFFFAOYSA-N
2-chloro-4-nitrophenyl isothiocyanate, 2-chloro-4-nitrophenylisothiocyanate, benzene,2-chloro-1-isothiocyanato-4-nitro, 2-chloro-4-nitrobenzenisothiocyanate, acmc-1cg9c, 2-chloranyl-1-isothiocyanato-4-nitro-benzene
2757736
2-chloro-1-isothiocyanato-4-nitrobenzene
[O-][N+](=O)C1=CC(Cl)=C(C=C1)N=C=S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 95°C to 99°C
Odor Irritating
Quantity 1 g
UN Number UN2923
Beilstein 2972987
Sensitivity Moisture sensitive
Formula Weight 214.63
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-nitrophenyl isothiocyanate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'