missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitrobenzonitrile, 97%, Maybridge

Brand:  Maybridge XAX00005EA

Code : 31

Additional Details : Weight : 0.01000kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Packaging Amber Glass Bottle
Quantity 10g

 View more versions of this product

Product Code. 10415413

  • £52.00 / 10g

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

28163-00-0
182.563
2-chloro-4-nitro-benzonitrile, benzonitrile, 2-chloro-4-nitro, 2-chloro-4-nitrobenzenecarbonitrile, pubchem18759, acmc-1ceg9, 2-chloro-4-nitrobenzonitril, chloronitrobenzenecarbonitrile, 3-chloro-4-cyanonitrobenzene, ksc497k1r
2-chloro-4-nitrobenzonitrile
C7H3ClN2O2
ZIGQFRQZYVQQDR-UHFFFAOYSA-N
141401
C1=CC(=C(C=C1[N+](=O)[O-])Cl)C#N

Specifications

2-chloro-4-nitrobenzonitrile
10g
97%
2-chloro-4-nitro-benzonitrile, benzonitrile, 2-chloro-4-nitro, 2-chloro-4-nitrobenzenecarbonitrile, pubchem18759, acmc-1ceg9, 2-chloro-4-nitrobenzonitril, chloronitrobenzenecarbonitrile, 3-chloro-4-cyanonitrobenzene, ksc497k1r
C1=CC(=C(C=C1[N+](=O)[O-])Cl)C#N
182.563
182.57
Amber Glass Bottle
28163-00-0
C7H3ClN2O2
ZIGQFRQZYVQQDR-UHFFFAOYSA-N
2-chloro-4-nitrobenzonitrile
141401
97%
Videos
SDS Documents