missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-4-nitrobenzoic acid, 98%

Brand:  Alfa Aesar™ A11602.22

Code : NEW

Additional Details : CAS Number : 99-60-5 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11484683

  • £24.00 / 100g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-4-nitrobenzoic acid
C7H4ClNO4
MFCD00007209
99-60-5
201.57
1912836

Specifications

2-Chloro-4-nitrobenzoic acid
100g
Odorless
C7H4ClNO4
MFCD00007209
140° to 144°C
99-60-5
98%
201.57
1912836
Videos
SDS