missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitro-N,N-di-n-propylaniline, 97%, Thermo Scientific™

Catalog Number p-7055360
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
6216-91-7
C12H17ClN2O2
256.73
OYJJPFCGGNTTTO-UHFFFAOYSA-N
2-chloro-4-nitro-nn-dipropylaniline, 4-nitro-2-chloro-n,n-di-n-propyl aniline, benzenamine,2-chloro-4-nitro-n,n-dipropyl, 2-chloro-4-nitrophenyl dipropylamine, 2-chloro-4-nitro-phenyl-dipropyl-amine
138688
2-chloro-4-nitro-N,N-dipropylaniline
CCCN(CCC)C1=C(C=C(C=C1)[N+](=O)[O-])Cl
This item is not returnable. View return policy
6216-91-7
C12H17ClN2O2
256.73
OYJJPFCGGNTTTO-UHFFFAOYSA-N
2-chloro-4-nitro-nn-dipropylaniline, 4-nitro-2-chloro-n,n-di-n-propyl aniline, benzenamine,2-chloro-4-nitro-n,n-dipropyl, 2-chloro-4-nitrophenyl dipropylamine, 2-chloro-4-nitro-phenyl-dipropyl-amine
138688
2-chloro-4-nitro-N,N-dipropylaniline
CCCN(CCC)C1=C(C=C(C=C1)[N+](=O)[O-])Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN2811
Formula Weight 256.73
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Chloro-4-nitro-N,N-di-n-propylaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'