missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-4-methylsulphonylaniline, 95%, ACROS Organics™

Brand:  Acros Organics 424990100

Code : 29

Additional Details : CAS Number : 13244-35-4 Weight : 0.06000kg


Quantity 10g
Packaging Glass bottle

 View more versions of this product

Product Code. 10187293

  • £170.60 / 10g


Estimated Shipment
16-03-2018
Add to basket


Specifications

Chemical Identifiers

2-chloro-4-methylsulphonylaniline
94.0
≥94%
C7H8ClNO2S
MFCD00152349
13244-35-4
100.0
95%
205.66

Specifications

2-chloro-4-methylsulphonylaniline
94.0
100.0
191° to 198°C
Pale purple to gray-brown
≥94%
C7H8ClNO2S
MFCD00152349
13244-35-4
Authentic
Glass bottle
10g
Crystalline Powder
95%
205.66
Videos
SDS