missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(methylsulfonyl)benzoic acid, 95%, Maybridge

£10.00 - £46.00

Chemical Identifiers

CAS 53250-83-2
Molecular Formula C8H7ClO4S
Molecular Weight (g/mol) 234.65
MDL Number MFCD00216496
InChI Key CTTWSFIIFMWHLQ-UHFFFAOYSA-N
Synonym 2-chloro-4-methylsulfonyl benzoic acid, 2-chloro-4-methylsulphonylbenzoic acid, 2-chloro-4-methanesulfonylbenzoic acid, benzoic acid, 2-chloro-4-methylsulfonyl, 2-chloro-4-methanesulfonyl-benzoic acid, 2-chloro-4-methylsulfonyl benzoic acid dry basis, maybridge1_004133, cmba, pubchem20281, acmc-1aupa
PubChem CID 735863
IUPAC Name 2-chloro-4-methylsulfonylbenzoic acid
SMILES CS(=O)(=O)C1=CC(=C(C=C1)C(=O)O)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10683625 SDS View Documents Maybridge
JFD01500EA
10g £10.00
10g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 23-03-2020
Please to purchase this item. Need a web account? Register with us today!
10265274 SDS View Documents Maybridge
JFD01500EB
25g £18.00
25g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 23-03-2020
Please to purchase this item. Need a web account? Register with us today!
10406794 SDS View Documents Maybridge
JFD01500FA
100g £46.00
100g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 23-03-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

53250-83-2
234.65
CTTWSFIIFMWHLQ-UHFFFAOYSA-N
735863
CS(=O)(=O)C1=CC(=C(C=C1)C(=O)O)Cl
C8H7ClO4S
MFCD00216496
2-chloro-4-methylsulfonyl benzoic acid, 2-chloro-4-methylsulphonylbenzoic acid, 2-chloro-4-methanesulfonylbenzoic acid, benzoic acid, 2-chloro-4-methylsulfonyl, 2-chloro-4-methanesulfonyl-benzoic acid, 2-chloro-4-methylsulfonyl benzoic acid dry basis, maybridge1_004133, cmba, pubchem20281, acmc-1aupa
2-chloro-4-methylsulfonylbenzoic acid

Specifications

2-Chloro-4-(methylsulfonyl)benzoic acid
10g
95.0
C8H7ClO4S
2-chloro-4-methylsulfonyl benzoic acid, 2-chloro-4-methylsulphonylbenzoic acid, 2-chloro-4-methanesulfonylbenzoic acid, benzoic acid, 2-chloro-4-methylsulfonyl, 2-chloro-4-methanesulfonyl-benzoic acid, 2-chloro-4-methylsulfonyl benzoic acid dry basis, maybridge1_004133, cmba, pubchem20281, acmc-1aupa
CS(=O)(=O)C1=CC(=C(C=C1)C(=O)O)Cl
234.65
234.66
Amber glass bottle
53250-83-2
95%
MFCD00216496
CTTWSFIIFMWHLQ-UHFFFAOYSA-N
2-chloro-4-methylsulfonylbenzoic acid
735863
95%