missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-(methylsulfonyl)benzoic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
100 g
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Quantity | 100 g |
Formula Weight | 234.66 |
Percent Purity | 95% |
Chemical Name or Material | 2-Chloro-4-(methylsulfonyl)benzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'