missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methoxypyrimidine, 98%, Thermo Scientific Chemicals

Catalog Number p-3770841
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
22536-63-6
C5H5ClN2O
144.56
MFCD00194055
BDXYNMVQMBCTDB-UHFFFAOYSA-N
2-chloro-4-methoxy-pyrimidine, pyrimidine, 2-chloro-4-methoxy, acmc-20a0n4, 2-chloro4-methoxypyrimidine, 4-methoxy-2-chloropyrimidine, ksc494k1r, 2-chloro-4-methoxypyrimidine, 2-chloro-4-methyloxy pyrimidine, 2-chloro-4-methoxy-1,2-diazine
588935
2-chloro-4-methoxypyrimidine
COC1=NC(=NC=C1)Cl
This item is not returnable. View return policy
22536-63-6
C5H5ClN2O
144.56
MFCD00194055
BDXYNMVQMBCTDB-UHFFFAOYSA-N
2-chloro-4-methoxy-pyrimidine, pyrimidine, 2-chloro-4-methoxy, acmc-20a0n4, 2-chloro4-methoxypyrimidine, 4-methoxy-2-chloropyrimidine, ksc494k1r, 2-chloro-4-methoxypyrimidine, 2-chloro-4-methyloxy pyrimidine, 2-chloro-4-methoxy-1,2-diazine
588935
2-chloro-4-methoxypyrimidine
COC1=NC(=NC=C1)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 54°C to 57°C
Color White to Yellow
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 144.56
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-methoxypyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-4-methoxypyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'