missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-4-iodo-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide, 95%

Brand:  Alfa Aesar™ H65550.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15499726

  • £59.78 / 1g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-4-iodo-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide
411.67
MFCD27975782

Specifications

2-Chloro-4-iodo-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide
1g
95%
MFCD27975782
Light Sensitive
Solid
411.67
Videos
SDS