missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-4-fluorophenol, 98+%

Brand:  Alfa Aesar™ A10864.06

Code : NEW

Additional Details : CAS Number : 1996-41-4 Weight : 0.00500kg Transport : UN number : 3265 Chem class : 8 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 11449852

  • £16.90 / 5g


Estimated Shipment date
from Supplier 25-05-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-4-fluorophenol
C6H4ClFO
MFCD00002168
1861277
1996-41-4
146.55
UN3265

Specifications

2-Chloro-4-fluorophenol
Phenol-like
20° to 23°C
1996-41-4
C6H4ClFO
MFCD00002168
1861277
75°C (167°F)
171° to 172°C
5g
≥98%
146.55
UN3265
Videos
SDS