missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorobenzoyl chloride, 97%, Thermo Scientific™

Catalog Number p-7027007
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
21900-54-9
C7H3Cl2FO
192.998
MFCD00084954
POIAZJJVWRVLBO-UHFFFAOYSA-N
2736546
2-chloro-4-fluorobenzoyl chloride
C1=CC(=C(C=C1F)Cl)C(=O)Cl
This item is not returnable. View return policy
21900-54-9
C7H3Cl2FO
192.998
MFCD00084954
POIAZJJVWRVLBO-UHFFFAOYSA-N
2736546
2-chloro-4-fluorobenzoyl chloride
C1=CC(=C(C=C1F)Cl)C(=O)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 82°C to 84°C (15 mmHg)
Refractive Index 1.551
Quantity 1 g
UN Number UN3265
Beilstein 2254071
Sensitivity Moisture sensitive
Synonym 2-chloro-4-fluorobenzoylchloride, 2-chloro-4-fluoro-benzoyl chloride, 2-chloro-4-fluorobenzene-1-carbonyl chloride, benzoyl chloride, 2-chloro-4-fluoro, benzoyl chloride,2-chloro-4-fluoro, acmc-20anzq, ksc547s3d, 2-chlor-4-fluorbenzoylchlorid, 2chloro-4-fluorobenzoyl chloride, 4-fluoro-2-chlorobenzoyl chloride
Formula Weight 193.01
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluorobenzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'