missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-fluorobenzoyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Boiling Point | 82°C to 84°C (15 mmHg) |
Refractive Index | 1.551 |
Quantity | 1 g |
UN Number | UN3265 |
Beilstein | 2254071 |
Sensitivity | Moisture sensitive |
Synonym | 2-chloro-4-fluorobenzoylchloride, 2-chloro-4-fluoro-benzoyl chloride, 2-chloro-4-fluorobenzene-1-carbonyl chloride, benzoyl chloride, 2-chloro-4-fluoro, benzoyl chloride,2-chloro-4-fluoro, acmc-20anzq, ksc547s3d, 2-chlor-4-fluorbenzoylchlorid, 2chloro-4-fluorobenzoyl chloride, 4-fluoro-2-chlorobenzoyl chloride |
Formula Weight | 193.01 |
Percent Purity | 97% |
Chemical Name or Material | 2-Chloro-4-fluorobenzoyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'