missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-chloro-4-fluorobenzene-1-sulfonyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Linear Formula | Cl(F)C6H3(SO2Cl) |
Quantity | 25 g |
Formula Weight | 229.06 |
Percent Purity | 97% |
Chemical Name or Material | 2-Chloro-4-fluorobenzene-1-sulfonyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'