missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-4,5-difluorobenzaldehyde, 98%

Brand:  Alfa Aesar™ H64843.06

Code : NEW

Additional Details : CAS Number : 165047-23-4 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15492229

  • £361.46 / 5g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-4,5-difluorobenzaldehyde
176.55
13767245
165047-23-4
MFCD07782050

Specifications

2-Chloro-4,5-difluorobenzaldehyde
165047-23-4
98%
MFCD07782050
5g
Solid
176.55
13767245
Videos
SDS