missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(4-morpholinyl)quinazoline, 97%, Thermo Scientific™

Catalog Number 15459648
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
39213-05-3
C12H12ClN3O
249.70
MFCD03083869
NJBDGZVAZRMIFM-UHFFFAOYSA-N
4-2-chloroquinazolin-4-yl morpholine, 2-chloro-4-morpholino-quinazoline, 2-chloro-4-morpholin-4-yl quinazoline, 2-chloro-4-morpholinoquinazoline, 2-chloro-4-4-morpholinyl quinazoline, 2-chloro-4-morpholin-4-yl-quinazoline
2338466
4-(2-chloroquinazolin-4-yl)morpholine
ClC1=NC(N2CCOCC2)=C2C=CC=CC2=N1
This item is not returnable. View return policy
39213-05-3
C12H12ClN3O
249.70
MFCD03083869
NJBDGZVAZRMIFM-UHFFFAOYSA-N
4-2-chloroquinazolin-4-yl morpholine, 2-chloro-4-morpholino-quinazoline, 2-chloro-4-morpholin-4-yl quinazoline, 2-chloro-4-morpholinoquinazoline, 2-chloro-4-4-morpholinyl quinazoline, 2-chloro-4-morpholin-4-yl-quinazoline
2338466
4-(2-chloroquinazolin-4-yl)morpholine
ClC1=NC(N2CCOCC2)=C2C=CC=CC2=N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 249.7
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Chloro-4-(4-morpholinyl)quinazoline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'