missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-4-(1-piperidinyl)quinazoline, 97%

Brand:  Alfa Aesar™ H63348.03

Code : NEW

Additional Details : CAS Number : 59870-45-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15479998

  • £122.96 / 1g


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-4-(1-piperidinyl)quinazoline
59870-45-0
247.72
MFCD03692973

Specifications

2-Chloro-4-(1-piperidinyl)quinazoline
59870-45-0
97%
MFCD03692973
1g
Solid
247.72
Videos
SDS