missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-3-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 36.0°C to 40.0°C |
Boiling Point | 166.0°C to 168.0°C |
Flash Point | 82°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Quantity | 5 g |
Solubility Information | Solubility in water: insoluble. Other solubilities: reacts at high temperature with strong acids and,alkalis |
Show More |
Safety and Handling
Product Identifier
- 2-Chloro-3-(trifluoromethyl)pyridine
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 1
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
- H314-Causes severe skin burns and eye damage.
- H311-Toxic in contact with skin.
- H412-Harmful to aquatic life with long lasting effects.
- H301-Toxic if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'