missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Catalog Number p-4228546
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
65753-47-1
C6H3ClF3N
181.54
RXATZPCCMYMPME-UHFFFAOYSA-N
2-chloro-3-trifluoromethyl pyridine, 2-chloro-3-trifluoro methylpyridine, pyridine, 2-chloro-3-trifluoromethyl, 2,3-ctf, 2-chloro-3-trifluoromethyl-pyridine, 3-trifluoromethyl-2-chloropyridine, zlchem 248, pubchem2399, acmc-1bejp, ksc352q1t
589833
2-chloro-3-(trifluoromethyl)pyridine
C1=CC(=C(N=C1)Cl)C(F)(F)F
This item is not returnable. View return policy
65753-47-1
C6H3ClF3N
181.54
RXATZPCCMYMPME-UHFFFAOYSA-N
2-chloro-3-trifluoromethyl pyridine, 2-chloro-3-trifluoro methylpyridine, pyridine, 2-chloro-3-trifluoromethyl, 2,3-ctf, 2-chloro-3-trifluoromethyl-pyridine, 3-trifluoromethyl-2-chloropyridine, zlchem 248, pubchem2399, acmc-1bejp, ksc352q1t
589833
2-chloro-3-(trifluoromethyl)pyridine
C1=CC(=C(N=C1)Cl)C(F)(F)F
CAS Min % 96.0
CAS Max % 100.0
Melting Point 36.0°C to 40.0°C
Boiling Point 166.0°C to 168.0°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: reacts at high temperature with strong acids and,alkalis
Formula Weight 181.54
Percent Purity 97%
Physical Form Crystals
Chemical Name or Material 2-chloro-3-(trifluoromethyl)pyridine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Chloro-3-(trifluoromethyl)pyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H412-Harmful to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'