missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-(trifluoromethyl)benzoic acid, 97+%, Thermo Scientific™

Catalog Number p-7026642
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
39226-97-6
C8H4ClF3O2
224.563
MFCD00792415
AXOAWWUSRZCGKS-UHFFFAOYSA-N
2-chloro-3-trifluoromethyl benzoic acid, 2-chloro-3-trifluoromethyl-benzoic acid, benzoic acid, 2-chloro-3-trifluoromethyl, 2-chloro-3-trifluoromethyl benzoicacid, acmc-1ahj6, ksc496e0b, rarechem al be 0475, rarechem al be 0520, rarechem al bo 0356
2736677
2-chloro-3-(trifluoromethyl)benzoic acid
C1=CC(=C(C(=C1)C(F)(F)F)Cl)C(=O)O
This item is not returnable. View return policy
39226-97-6
C8H4ClF3O2
224.563
MFCD00792415
AXOAWWUSRZCGKS-UHFFFAOYSA-N
2-chloro-3-trifluoromethyl benzoic acid, 2-chloro-3-trifluoromethyl-benzoic acid, benzoic acid, 2-chloro-3-trifluoromethyl, 2-chloro-3-trifluoromethyl benzoicacid, acmc-1ahj6, ksc496e0b, rarechem al be 0475, rarechem al be 0520, rarechem al bo 0356
2736677
2-chloro-3-(trifluoromethyl)benzoic acid
C1=CC(=C(C(=C1)C(F)(F)F)Cl)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 132°C to 136°C
Quantity 1 g
Formula Weight 224.57
Percent Purity ≥97%
Chemical Name or Material 2-Chloro-3-(trifluoromethyl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'