missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-3-pyridinemethanol, 97%

Brand:  Alfa Aesar™ H63341.03

Code : NEW

Additional Details : CAS Number : 42330-59-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449988

  • £27.20 / 1g


Estimated Shipment date
from Supplier 23-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-3-pyridinemethanol
143.57
1525378
42330-59-6
MFCD01935961
2-Chloro-3-(hydroxymethyl)pyridine

Specifications

2-Chloro-3-pyridinemethanol
61° to 65°C (dec.)
42330-59-6
97%
MFCD01935961
2-Chloro-3-(hydroxymethyl)pyridine
112°C (0.2mmHg)
1g
Solid
143.57
1525378
Videos
SDS