missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-3-nitrothiophene, 97%, Thermo Scientific™

Catalog Number p-2698038
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
5330-98-3
C4H2ClNO2S
163.58
MFCD00052159
OFPRGOSJWUNETN-UHFFFAOYSA-N
thiophene,2-chloro-3-nitro, thiophene, 2-chloro-3-nitro, 2-chloro-3-nitro-thiophene, pubchem5466
220186
2-chloro-3-nitrothiophene
[O-][N+](=O)C1=C(Cl)SC=C1
This item is not returnable. View return policy
5330-98-3
C4H2ClNO2S
163.58
MFCD00052159
OFPRGOSJWUNETN-UHFFFAOYSA-N
thiophene,2-chloro-3-nitro, thiophene, 2-chloro-3-nitro, 2-chloro-3-nitro-thiophene, pubchem5466
220186
2-chloro-3-nitrothiophene
[O-][N+](=O)C1=C(Cl)SC=C1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 163.58
Percent Purity 97%
Chemical Name or Material 2-chloro-3-nitrothiophene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'