missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-nitropyridine, 99%, Thermo Scientific Chemicals

Catalog Number p-7081926
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
5470-18-8
C5H3ClN2O2
158.541
MFCD00006232
UUOLETYDNTVQDY-UHFFFAOYSA-N
pyridine, 2-chloro-3-nitro, 3-nitro-2-chloropyridine, 2-chloro-3-nitro-pyridine, 2-chloro-3-nitro pyridine, 2-chlor-3-nitropyridin, zlchem 303, pubchem1229, 2-chloro-nitropyridine, 2-chloro-3nitropyridine, 2-chloro3-nitropyridine
79613
2-chloro-3-nitropyridine
C1=CC(=C(N=C1)Cl)[N+](=O)[O-]
This item is not returnable. View return policy
5470-18-8
C5H3ClN2O2
158.541
MFCD00006232
UUOLETYDNTVQDY-UHFFFAOYSA-N
pyridine, 2-chloro-3-nitro, 3-nitro-2-chloropyridine, 2-chloro-3-nitro-pyridine, 2-chloro-3-nitro pyridine, 2-chlor-3-nitropyridin, zlchem 303, pubchem1229, 2-chloro-nitropyridine, 2-chloro-3nitropyridine, 2-chloro3-nitropyridine
79613
2-chloro-3-nitropyridine
C1=CC(=C(N=C1)Cl)[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 99°C to 103°C
Flash Point 185°C (365°F)
Quantity 5 g
Beilstein 124482
Formula Weight 158.54
Percent Purity 99%
Chemical Name or Material 2-Chloro-3-nitropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'