missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-methylpyrazine, 95%, Thermo Scientific™

Catalog Number 15408218
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
95-58-9
C5H5ClN2
128.559
MFCD00023279
WZHWPZQQPWKEAV-UHFFFAOYSA-N
pyrazine, 2-chloro-3-methyl, 2-methyl-3-chloropyrazine, 2-chloro-3-methyl-pyrazine, 2-chloro-3-methyl pyrazine, pubchem2149, 2-chloro-3-methylpyrazin, 3-chloro-2-methylpyrazine, 3-chloro-2-methyl pyrazine, wzhwpzqqpwkeav-uhfffaoysa, 2-chloro-3, 5 , 6-ethylpyrazine
66769
2-chloro-3-methylpyrazine
CC1=NC=CN=C1Cl
This item is not returnable. View return policy
95-58-9
C5H5ClN2
128.559
MFCD00023279
WZHWPZQQPWKEAV-UHFFFAOYSA-N
pyrazine, 2-chloro-3-methyl, 2-methyl-3-chloropyrazine, 2-chloro-3-methyl-pyrazine, 2-chloro-3-methyl pyrazine, pubchem2149, 2-chloro-3-methylpyrazin, 3-chloro-2-methylpyrazine, 3-chloro-2-methyl pyrazine, wzhwpzqqpwkeav-uhfffaoysa, 2-chloro-3, 5 , 6-ethylpyrazine
66769
2-chloro-3-methylpyrazine
CC1=NC=CN=C1Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Quantity 250 mg
Formula Weight 128.56
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Chloro-3-methylpyrazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'