missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-fluoropyridine-6-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog Number 15459116
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
860296-24-8
C6H3ClFNO2
175.543
MFCD13185819
JHFKXHUKNYFWJM-UHFFFAOYSA-N
6-chloro-5-fluoropicolinic acid, 2-chloro-3-fluoropyridine-6-carboxylicacid, 2-chloro-3-fluoropyridine-6-carboxylic acid, 6-chloro-5-fluoro-pyridine-2-carboxylic acid, 2-chloro-3-fluoro-6-carboxypyridine, 2-chloro-3-fluoro-6-pyridinecarboxylic acid
11412672
6-chloro-5-fluoropyridine-2-carboxylic acid
C1=CC(=NC(=C1F)Cl)C(=O)O
This item is not returnable. View return policy
860296-24-8
C6H3ClFNO2
175.543
MFCD13185819
JHFKXHUKNYFWJM-UHFFFAOYSA-N
6-chloro-5-fluoropicolinic acid, 2-chloro-3-fluoropyridine-6-carboxylicacid, 2-chloro-3-fluoropyridine-6-carboxylic acid, 6-chloro-5-fluoro-pyridine-2-carboxylic acid, 2-chloro-3-fluoro-6-carboxypyridine, 2-chloro-3-fluoro-6-pyridinecarboxylic acid
11412672
6-chloro-5-fluoropyridine-2-carboxylic acid
C1=CC(=NC(=C1F)Cl)C(=O)O

It is an important intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Beilstein 10063182
Solubility Information Slightly soluble in water.
Formula Weight 175.54
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Chloro-3-fluoropyridine-6-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'