missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-3-fluoro-5-methylpyridine, 95%

Brand:  Alfa Aesar™ H61972.03

Code : NEW

Additional Details : CAS Number : 34552-15-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15428568

  • £43.60 / 1g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-3-fluoro-5-methylpyridine
34552-15-3
145.56
MFCD06658238

Specifications

2-Chloro-3-fluoro-5-methylpyridine
1g
Solid
145.56
White
34552-15-3
95%
MFCD06658238
Videos
SDS