missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-3-fluoro-4-iodopyridine, 98%

Brand:  Alfa Aesar™ H64610.06

Code : NEW

Additional Details : CAS Number : 148639-07-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15481889

  • £298.92 / 5g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-3-fluoro-4-iodopyridine
257.43
5862293
148639-07-0
MFCD04972389

Specifications

2-Chloro-3-fluoro-4-iodopyridine
104°C
148639-07-0
98%
MFCD04972389
Light Sensitive
5g
Solid
257.43
5862293
Videos
SDS