missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-ethylbenzoxazolium tetrafluoroborate, 97%, ACROS Organics™

Brand:  Acros Organics 214000010

Code : 29

Additional Details : CAS Number : 63212-53-3 Weight : 0.05100kg Transport : UN number : 1759 Chem class : 8 Pack group : III


Quantity 1g

 View more versions of this product

Product Code. 10135080

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

2-Chloro-3-ethylbenzoxazolium tetrafluoroborate
63212-53-3
100.0
97%
269.42
97%
96.0
≥96.0%
C9H9ClNO·BF4
MFCD00011943

Specifications

2-Chloro-3-ethylbenzoxazolium tetrafluoroborate
1g
63212-53-3
100.0
Glass bottle
≥96.0%
C9H9ClNO·BF4
MFCD00011943
Beige
97%
96.0
Authentic
Crystals or Crystalline Powder
97%
269.42
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Chloro-3-ethylbenzoxazolium tetrafluoroborate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS