missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-3,6-difluorobenzoic acid, 97%

£19.00 - £67.70

Chemical Identifiers

CAS 287172-74-1
Molecular Formula C7H3ClF2O2
Molecular Weight (g/mol) 192.546
MDL Number MFCD01631441
InChI Key COWJYTLVSFKOPM-UHFFFAOYSA-N
Synonym 2-chloro-3,6-difluoro-benzoic acid, benzoic acid, 2-chloro-3,6-difluoro, pubchem8530, acmc-1cr2e, benzoic acid,2-chloro-3,6-difluoro, 2-chloro-3,6-difluorobenzoic acid
PubChem CID 2773530
IUPAC Name 2-chloro-3,6-difluorobenzoic acid
SMILES C1=CC(=C(C(=C1F)C(=O)O)Cl)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11461107 SDS View Documents Alfa Aesar™
B24105.03
1g £19.00
1g View alternative products
Save Up to 
»
EU Stock 2 Please to purchase this item. Need a web account? Register with us today!
11471107 SDS View Documents Alfa Aesar™
B24105.06
5g £67.70
5g View alternative products
Save Up to 
»
EU Stock 1 Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

287172-74-1
192.546
COWJYTLVSFKOPM-UHFFFAOYSA-N
2773530
C1=CC(=C(C(=C1F)C(=O)O)Cl)F
C7H3ClF2O2
MFCD01631441
2-chloro-3,6-difluoro-benzoic acid, benzoic acid, 2-chloro-3,6-difluoro, pubchem8530, acmc-1cr2e, benzoic acid,2-chloro-3,6-difluoro, 2-chloro-3,6-difluorobenzoic acid
2-chloro-3,6-difluorobenzoic acid

Specifications

2-Chloro-3,6-difluorobenzoic acid
1g
C7H3ClF2O2
2-chloro-3,6-difluoro-benzoic acid, benzoic acid, 2-chloro-3,6-difluoro, pubchem8530, acmc-1cr2e, benzoic acid,2-chloro-3,6-difluoro, 2-chloro-3,6-difluorobenzoic acid
C1=CC(=C(C(=C1F)C(=O)O)Cl)F
192.546
192.55
126°C to 128°C
287172-74-1
MFCD01631441
COWJYTLVSFKOPM-UHFFFAOYSA-N
2-chloro-3,6-difluorobenzoic acid
2773530
97%