missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-3-(5-methyl-1,2,4-oxadiazol-3-yl)pyridine, 97%

Brand:  Alfa Aesar™ H65238.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15499486

  • £88.93 / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-3-(5-methyl-1,2,4-oxadiazol-3-yl)pyridine
195.61
MFCD08273493

Specifications

2-Chloro-3-(5-methyl-1,2,4-oxadiazol-3-yl)pyridine
97%
1g
MFCD08273493
Solid
White to pale yellow
195.61
Videos
SDS