missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-3,5-bis(trifluoromethyl)aniline, 98%

Brand:  Alfa Aesar™ H64119.14

Code : NEW

Additional Details : CAS Number : 201593-90-0 Weight : 0.02500kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 15401179

  • £76.45 / 25g

Estimated Shipment: 23-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
2-Chloro-3,5-bis(trifluoromethyl)aniline is used as dyestuff Intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

201593-90-0
263.567
JKUFETFGEJPHEM-UHFFFAOYSA-N
2757429
C1=C(C=C(C(=C1C(F)(F)F)Cl)N)C(F)(F)F
C8H4ClF6N
MFCD04038716
2-chloro-3,5-bis trifluoromethyl aniline, 3,5-bis trifluoromethyl-2-chloroaniline, 2-chloro-3,5-bis-trifluoromethyl aniline, benzenamine, 2-chloro-3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl-2-chlorophenylamine, pubchem2938, 2-chloro-3,5-bis-trifluoromethyl-aniline, 3,5-bis trifluoromethyl-2-chloro aniline, 2-chloranyl-3,5-bis trifluoromethyl aniline, 2-chloro-3,5-bis-trifluoromethyl-phenylamine
2-chloro-3,5-bis(trifluoromethyl)aniline

Specifications

2-Chloro-3,5-bis(trifluoromethyl)aniline
1.459
25g
C8H4ClF6N
UN2810
2-chloro-3,5-bis trifluoromethyl aniline, 3,5-bis trifluoromethyl-2-chloroaniline, 2-chloro-3,5-bis-trifluoromethyl aniline, benzenamine, 2-chloro-3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl-2-chlorophenylamine, pubchem2938, 2-chloro-3,5-bis-trifluoromethyl-aniline, 3,5-bis trifluoromethyl-2-chloro aniline, 2-chloranyl-3,5-bis trifluoromethyl aniline, 2-chloro-3,5-bis-trifluoromethyl-phenylamine
JKUFETFGEJPHEM-UHFFFAOYSA-N
2-chloro-3,5-bis(trifluoromethyl)aniline
2757429
Liquid
1.595 g/mL
Colorless to Yellow
201593-90-0
MFCD04038716
13730810
Slightly soluble in water.
C1=C(C=C(C(=C1C(F)(F)F)Cl)N)C(F)(F)F
263.567
263.57
98%
Videos
SDS Documents
Certificates