missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2-ethoxyacetic acid ethyl ester, 90%, remainder mainly ethyl diethoxyacetat, Thermo Scientific Chemicals

Catalog Number p-3211630
Click to view available options
Quantity:
50 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
34006-60-5
C6H11ClO3
166.60
MFCD00043953
WDCHMODANTVRTF-UHFFFAOYNA-N
2-chloro-2-ethoxyacetic acid ethyl ester, ethyl 2-chloro-2-ethoxy-acetate, ethyl2-chloro-2-ethoxyacetate, ethyl chloro ethoxy acetate, chloro ethoxy acetic acid ethyl ester, ethyl alpha-chloro-alpha-ethoxyacetate, ethyl alpha-chloro-alpha-ethoxy-acetate, 2-chloro-2-ethoxy-acetic acid ethyl ester, acetic acid,2-chloro-2-ethoxy-, ethyl ester
2724873
ethyl 2-(1-chloroethoxy)acetate
CCOC(=O)COC(C)Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
34006-60-5
C6H11ClO3
166.60
MFCD00043953
WDCHMODANTVRTF-UHFFFAOYNA-N
2-chloro-2-ethoxyacetic acid ethyl ester, ethyl 2-chloro-2-ethoxy-acetate, ethyl2-chloro-2-ethoxyacetate, ethyl chloro ethoxy acetate, chloro ethoxy acetic acid ethyl ester, ethyl alpha-chloro-alpha-ethoxyacetate, ethyl alpha-chloro-alpha-ethoxy-acetate, 2-chloro-2-ethoxy-acetic acid ethyl ester, acetic acid,2-chloro-2-ethoxy-, ethyl ester
2724873
ethyl 2-(1-chloroethoxy)acetate
CCOC(=O)COC(C)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 88.0
CAS Max % 100.0
Color Brown-Orange to Colorless
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula C2H5O(Cl)CHC(O)OC2H5
Refractive Index 1.4307
Quantity 50 g
Formula Weight 166.6
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 2-Chloro-2-ethoxyacetic acid ethyl ester
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-2-ethoxyacetic acid ethyl ester, remainder mainly ethyl diethoxyacetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'