missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chloro-1-tritylbenzimidazole, 97%

Brand:  Alfa Aesar™ H63743.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15470589

  • £129.32 / 5g


Estimated Shipment date
from Supplier 05-07-2018
Add to basket

Specifications

Chemical Identifiers

2-Chloro-1-tritylbenzimidazole
394.9
MFCD26143385

Specifications

2-Chloro-1-tritylbenzimidazole
5g
97%
MFCD26143385
White to yellow
Solid
394.9
Videos
SDS