missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Chloro-1-phenylethanol, 97%, Thermo Scientific Chemicals

Catalog Number 11383576
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1674-30-2
C8H9ClO
156.609
MFCD01075658
XWCQSILTDPAWDP-UHFFFAOYSA-N
styrene chlorohydrin, 1-phenyl-2-chloroethanol, +/--2-chloro-1-phenylethanol, 2-chloro-1-phenyl-ethanol, benzenemethanol, .alpha.-chloromethyl, benzenemethanol, a-chloromethyl, acmc-20apgz, benzyl alcohol, .alpha.-chloromethyl, acmc-20a5xf, 2-chloro-l-phenylethanol
92898
2-chloro-1-phenylethanol
C1=CC=C(C=C1)C(CCl)O
This item is not returnable. View return policy
1674-30-2
C8H9ClO
156.609
MFCD01075658
XWCQSILTDPAWDP-UHFFFAOYSA-N
styrene chlorohydrin, 1-phenyl-2-chloroethanol, +/--2-chloro-1-phenylethanol, 2-chloro-1-phenyl-ethanol, benzenemethanol, .alpha.-chloromethyl, benzenemethanol, a-chloromethyl, acmc-20apgz, benzyl alcohol, .alpha.-chloromethyl, acmc-20a5xf, 2-chloro-l-phenylethanol
92898
2-chloro-1-phenylethanol
C1=CC=C(C=C1)C(CCl)O

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Density 1.193
Boiling Point 110°C to 112°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.55
Quantity 5 g
UN Number UN2810
Beilstein 907322
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 156.61
Percent Purity 97%
Chemical Name or Material (+/-)-2-Chloro-1-phenylethanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'