missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,4-benzoquinone, 95%, Thermo Scientific Chemicals

Catalog Number 10653892
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
695-99-8
C6H3ClO2
142.54
MFCD00075260
WOGWYSWDBYCVDY-UHFFFAOYSA-N
2-chloro-1,4-benzoquinone, chloroquinone, monochloroquinone, 2-chloroquinone, chloro-p-benzoquinone, 2-chloro-p-benzoquinone, monochloro-p-benzoquinone, chloro-1,4-benzoquinone, p-benzoquinone, 2-chloro, 2,5-cyclohexadiene-1,4-dione, 2-chloro
69671
2-chlorocyclohexa-2,5-diene-1,4-dione
ClC1=CC(=O)C=CC1=O
This item is not returnable. View return policy
695-99-8
C6H3ClO2
142.54
MFCD00075260
WOGWYSWDBYCVDY-UHFFFAOYSA-N
2-chloro-1,4-benzoquinone, chloroquinone, monochloroquinone, 2-chloroquinone, chloro-p-benzoquinone, 2-chloro-p-benzoquinone, monochloro-p-benzoquinone, chloro-1,4-benzoquinone, p-benzoquinone, 2-chloro, 2,5-cyclohexadiene-1,4-dione, 2-chloro
69671
2-chlorocyclohexa-2,5-diene-1,4-dione
ClC1=CC(=O)C=CC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 52.0°C to 57.0°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 5 g
Formula Weight 142.54
Percent Purity 95%
Physical Form Crystals
Chemical Name or Material 2-Chloro-1, 4-benzoquinone, 95%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'