missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-1,3-thiazole, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 10 g |
Formula Weight | 119.57 |
Percent Purity | 97% |
Chemical Name or Material | 2-Chloro-1, 3-thiazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'