missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3-thiazole-5-carbaldehyde, 97%, Maybridge

Brand:  Maybridge CC62904DA

Code : 31

Additional Details : CAS Number : 95453-58-0 Weight : 0.05100kg

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10555164

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2-Chloro-1, 3-thiazole-5-carbaldehyde
96.5
97%
C4H2ClNOS
MFCD01568717
95453-58-0
100.0
97%
147.58

Specifications

2-Chloro-1, 3-thiazole-5-carbaldehyde
95453-58-0
100.0
97%
C4H2ClNOS
MFCD01568717
1g
96.5
Amber Glass Bottle
97%
147.58
Videos
SDS