missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3-dimethylimidazolinium chloride, 90%, Thermo Scientific Chemicals

Catalog Number p-4229140
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
37091-73-9
C5H10Cl2N2
169.05
MFCD09039290
AEBBXVHGVADBHA-UHFFFAOYSA-M
2-chloro-1,3-dimethylimidazolinium chloride, unii-56ka2a1u1d, 2-chloro-1,3-dimethyl-4,5-dihydro-1h-imidazol-3-ium chloride, dmc mi, 2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium chloride, pubchem16443, 2-chloro-1,3-dimethyl-4,5-dihydro-3h-imidazol-1-ium chloride, 1h-imidazolium, 2-chloro-4,5-dihydro-1,3-dimethyl-, chloride 1:1, 2-chloro-1,3-dimethylimidazolidin-2-ylium chloride
10176306
2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium;chloride
[Cl-].CN1CC[N+](C)=C1Cl
This item is not returnable. View return policy
37091-73-9
C5H10Cl2N2
169.05
MFCD09039290
AEBBXVHGVADBHA-UHFFFAOYSA-M
2-chloro-1,3-dimethylimidazolinium chloride, unii-56ka2a1u1d, 2-chloro-1,3-dimethyl-4,5-dihydro-1h-imidazol-3-ium chloride, dmc mi, 2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium chloride, pubchem16443, 2-chloro-1,3-dimethyl-4,5-dihydro-3h-imidazol-1-ium chloride, 1h-imidazolium, 2-chloro-4,5-dihydro-1,3-dimethyl-, chloride 1:1, 2-chloro-1,3-dimethylimidazolidin-2-ylium chloride
10176306
2-chloro-1,3-dimethyl-4,5-dihydroimidazol-1-ium;chloride
[Cl-].CN1CC[N+](C)=C1Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY
CAS Min % 88.0
CAS Max % 100.0
Melting Point 112°C to 121°C
Color White
Elemental Analysis 31.26 to 39.79% C, 14.58 to 18.56% N
Infrared Spectrum Authentic
Assay Percent Range 88% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15,3434
Formula Weight 169.05
Percent Purity 90%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-chloro-1, 3-dimethylimidazolinium chloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-1,3-dimethylimidazolinium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'