missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,1,1-triethoxyethane, 96%, ACROS Organics™

Brand:  Acros Organics 277670010

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 51076-95-0 Weight : 0.05100kg Transport : UN number : 1993 Chem class : 1 Pack group : II

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10370684

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications Description & Specifications

Chemical Identifiers

2-Chloro-1, 1, 1-triethoxyethane
51076-95-0
100.0
96%
(C2H5O)3CCH2Cl
MFCD00142889
96%
95.0
≥95%
C8H17ClO3
196.67

Specifications

2-Chloro-1, 1, 1-triethoxyethane
1g
51076-95-0
100.0
Glass bottle
Liquid
96%
(C2H5O)3CCH2Cl
MFCD00142889
Clear colorless
96%
95.0
Authentic
1.4215 to 1.4235 (20°C, 589nm)
≥95%
C8H17ClO3
196.67
Documentation

Technical Documents

Alternative Products